ContactUsLinkComponent
Interleukin-6 Receptor beta, CD130, Human ELISA, BioVendor
Interleukin-6 Receptor beta, CD130, Human ELISA, BioVendor
Catalog # 102552-438
Supplier:  BioVendor
CAS Number:  
undefined
Interleukin-6 Receptor beta, CD130, Human ELISA, BioVendor
Catalog # 102552-438
Supplier:  BioVendor
Supplier Number:  RGP008R
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Interleukin-6 Receptor beta (CD130) Human ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-438
  • Supplier no.:
    RGP008R
Specifications
About this item
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms.
  • Calibration Range: 56.25–1800 pg/mL
  • Limit of Detection: 15 pg/mL
  • Cell culture supernatant, Plasma, Serum: 100 uL/well
Interleukin-6 is a multi-functional cytokine that regulates immune responses, acute phase reactions and hematopoiesis, and it may play a central role in host defense mechanisms. IL-6 is usually not produced constitutively by normal cells, but its expression is readily induced by a variety of cytokines, lipopolysaccharide or viral infections.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.