Order Entry
United States
ContactUsLinkComponent
Interleukin-2, IL-2, Rat ELISA, BioVendor
Interleukin-2, IL-2, Rat ELISA, BioVendor
Catalog # 102552-360
Supplier:  BioVendor
CAS Number:  
undefined
Interleukin-2, IL-2, Rat ELISA, BioVendor
Catalog # 102552-360
Supplier:  BioVendor
Supplier Number:  RAF054R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    IL-2
  • Description:
    Interleukin-2 (IL-2) Rat ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-360
  • Supplier no.:
    RAF054R

Specifications

About this item

Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens.

  • Calibration Range: 62.5-2000 pg/mL
  • Limit of Detection: 11 pg/mL
  • Body fluids, Cell culture supernatant, Serum: 50 uL/well

Interleukin-2 (IL-2) plays a central role in the activation and proliferation of lymphocytes that have been primed by antigens. IL-2 plays a pivotal role in for the expansion of most T-cells, natural killer cells and B-cells during certain phases of their response. IL-2 gene expression is regulated at the transcriptional level by several activation pathways. Antigen-specific proliferation of helper and cytotoxic T-lymphocytes following stimulation is critically dependent on IL-2 expression, secretion, and binding to receptors for IL-2 induced in an autocrine fashion on the surface of T-cells. Apart from its most important role to mediate antigen-specific T-lymphocyte proliferation, IL-2 modulates the expression of IFN-g and major histocompatibility antigens, stimulates proliferation and differentiation of activated B-cells, augments natural killer cell activity and inhibits granulocyte-macrophage colony formation. Alterations in the ability of T-cells to synthesize IL-2 have been observed in physiologic and pathologic states.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.