Order Entry
United States
ContactUsLinkComponent
 
GM-CSF Mouse ELISA, BioVendor
Catalog # 102552-252
Supplier:  BioVendor
CAS Number:  
undefined
GM-CSF Mouse ELISA, BioVendor
Catalog # 102552-252
Supplier:  BioVendor
Supplier Number:  RAF017R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    GM-CSF
  • Description:
    GM-CSF Mouse ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-252
  • Supplier no.:
    RAF017R

Specifications

About this item

GM-CSF is a small glycoprotein growth factor which stimulates the production and function of neutrophils, eosinophils and monocytes

  • Calibration Range: 3.9-250 pg/mL
  • Limit of Detection: 1.98 pg/mL
  • Cell culture supernatant, Serum: 50 uL/well

GM-CSF is a small glycoprotein growth factor which stimulates the production and function of neutrophils, eosinophils and monocytes. It can be produced by a wide variety of tissue types. The biological effects of GM-CSF are mediated through its binding to cell surface receptors, which appear to be widely expressed by thermatopoietic cells and also by some non-thermatopoietic cells, such as endothelial cells.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.