ContactUsLinkComponent
Ghrelin Unacylated Mouse/Rat Express ELISA, BioVendor
Ghrelin Unacylated Mouse/Rat Express ELISA, BioVendor
Catalog # 102552-242
Supplier:  BioVendor
CAS Number:  
undefined
Ghrelin Unacylated Mouse/Rat Express ELISA, BioVendor
Catalog # 102552-242
Supplier:  BioVendor
Supplier Number:  RA394063400R
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Mouse
  • Description:
    Ghrelin Unacylated Mouse/Rat Express ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-242
  • Supplier no.:
    RA394063400R
Specifications
About this item
Ghrelin stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis.
  • Calibration Range: 1.96-250 pg/mL
  • Limit of Detection: 0.6 pg/mL
  • Plasma-EDTA: 10uL/well
Ghrelin, an endogenous ligand for the growth hormone secretagogue receptor, is synthesized principally in the stomach
Ghrelin, an endogenous ligand for the growth hormone secretagogue receptor, is synthesized principally in the stomach. It stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis. The peptide consists of 28 amino acids, with a n-octanoylation of the serine-3 residue, which is necessary for the biological activity mentioned below. Ghrelin is present in the peripheral circulation under two forms: acylated and unacylated.
It stimulates food intake and transduces signals to hypothalamic regulatory nuclei that controls energy homeostasis. The peptide consists of 28 amino acids, with a n-octanoylation of the serine-3 residue, which is necessary for the biological activity mentioned below. Ghrelin is present in the peripheral circulation under two forms: acylated and unacylated.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.