Orders LinkContactUsLinkComponent
EC Medium, Modified, BD Biosciences
 :   BD
EC Medium, Modified, BD Biosciences
 :  BD
EC Medium, Modified, BD Biosciences
Modified EC medium is used with novobiocin for the selective pre-enrichment of Escherichia coli.
  • Sodium chloride maintains osmotic balance of the medium
  • Novobiocin suppresses the growth of nuisance organisms
Lactose is the carbon source for EC medium, and bile salts mixtures are used to inhibit some Gram-positive cocci and sporeformers.
Product Family Options
  • Description
    Size
  • EC Medium, Modified
    500 g
     : -
  • EC Medium, Modified
    10 kg
    Each (10kg)
    Each (10kg) - $1,782.00
    $1,782.00
    Each (10kg)
       $1,782.00
     : 234001