Anti-CLEC14A Sheep Polyclonal Antibody
Catalog # 103244-046
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:CLEC14A
- Antigen Symbol:CLEC14A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:100 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose and Mannitol. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human CLEC14A Glu22-Ala397 Accession # Q86T13
- Cat. No.:103244-046
- Supplier no.:AF4968
Specifications
About this item
Type: Primary Antibodies
Antigen: CLEC14A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human