Anti-LYVE1 Rat Monoclonal Antibody (Allophycocyanin) [clone: 223322]
Catalog # 103239-342
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:LYVE-1
- Antigen Symbol:LYVE1
- Clonality:Monoclonal
- Clone:223322
- Conjugation:APC (Allophycocyanin)
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 Tests
- Storage Buffer:Supplied in a saline solution containing BSA and Sodium Azide.
- Storage Temperature:Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
- Immunogen:BaF/3 mouse pro-B cell line transfected with mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:103239-342
- Supplier no.:FAB2125A
Specifications
About this item
Type: Primary Antibodies
Antigen: LYVE1
Clonality: Monoclonal
Clone: 223322
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse