Anti-SSTR2 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 402038]
103235-366
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Somatostatin Receptor 2
- Antigen Symbol:SSTR2
- Clonality:Monoclonal
- Clone:402038
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 Tests
- Environmentally Preferable:
- Antigen Synonyms:SSTR2|SSR2
- Storage Buffer:Supplied in a saline solution containing BSA and Sodium Azide.
- Storage Temperature:Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
- Immunogen:NS0 mouse myeloma cell line transfected with Somatostatin R2/SSTR2 Met1-Ile369 Accession # P30874
- Cat. No.:103235-366
- Supplier no.:FAB4224P
Type: Primary Antibodies
Antigen: SSTR2
Clonality: Monoclonal
Clone: 402038
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human