Anti-TGM2 Sheep Polyclonal Antibody
103230-288
- Antibody Type:Primary
- Antigen Name:Transglutaminase 2/TGM2
- Antigen Symbol:TGM2
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Rat,Mouse
- Size:25 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Transglutaminase 2/TGM2 Ala2-Ala686 Accession # P21981
- Purification:Immunogen affinity purified
- Cat. No.:103230-288
- Supplier no.:AF5418-SP
Type: Primary Antibodies
Antigen: TGM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse, Rat