Anti-CLEC3B Sheep Polyclonal Antibody
103230-006
- Antibody Type:Primary
- Antigen Name:CLEC3B/Tetranectin
- Antigen Symbol:CLEC3B
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:100 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452
- Purification:Immunogen affinity purified
- Cat. No.:103230-006
- Supplier no.:AF5170
Type: Primary Antibodies
Antigen: CLEC3B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human