Anti-NINJ1 Sheep Polyclonal Antibody
Catalog # 103229-862
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Ninjurin 1
- Antigen Symbol:NINJ1
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:25 ug
- Antigen Synonyms:NINJURIN|NIN1|Nerve injury-induced protein 1|Ninjurin-1
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:E. coli-derived recombinant human Ninjurin-1 Asp2-Val81 Accession # Q92982
- Purification:Immunogen affinity purified
- Cat. No.:103229-862
- Supplier no.:AF5105-SP
Specifications
About this item
Type: Primary Antibodies
Antigen: NINJ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human