Anti-LAMP2 Goat Polyclonal Antibody
103229-662
- Antibody Type:Primary
- Antigen Name:LAMP-2/CD107b
- Antigen Symbol:LAMP2
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:25 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human LAMP2/CD107b Leu29-Phe375 Accession # P13473
- Purification:Immunogen affinity purified
- Cat. No.:103229-662
- Supplier no.:AF6228-SP
Type: Primary Antibodies
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human