Anti-APOH Goat Polyclonal Antibody
Catalog # 103229-118
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein H
- Antigen Symbol:APOH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:100 ug
- Antigen Synonyms:B2GPI|APC inhibitor|2GPI|BG|APOH|B2G1|activated protein C-binding protein|beta-2-glycoprotein I|beta-2-glycoprotein 1|anticardiolipin cofactor|beta|B2GP1|apo-H
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:E. coli-derived recombinant human Apolipoprotein H/ApoH Gly20-Cys345 Accession # P02749
- Purification:Immunogen affinity purified
- Cat. No.:103229-118
- Supplier no.:AF5087
Specifications
About this item
Type: Primary Antibodies
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human