Anti-CDSN Sheep Polyclonal Antibody
103228-246
- Antibody Type:Primary
- Antigen Name:Corneodesmosin
- Antigen Symbol:CDSN
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:100 ug
- Antigen Synonyms:HYPT2|HTSS|S|PSS1|PSS|HTSS1
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human Corneodesmosin Lys33-Pro529 (Ser153 del) Accession # NP_001255
- Purification:Immunogen affinity purified
- Cat. No.:103228-246
- Supplier no.:AF5725
Type: Primary Antibodies
Antigen: CDSN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human