Anti-PCDHA1 Sheep Polyclonal Antibody
103228-124
- Antibody Type:Primary
- Antigen Name:Protocadherin alpha 1
- Antigen Symbol:PCDHA1
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Size:25 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Protocadherin α1 Gln30-Pro689 Accession # Q9Y5I3
- Purification:Immunogen affinity purified
- Cat. No.:103228-124
- Supplier no.:AF5064-SP
Type: Primary Antibodies
Antigen: PCDHA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human