Order Entry
United States
ContactUsLinkComponent
Anti-FABP3 Sheep Polyclonal Antibody (Biotin)
Anti-FABP3 Sheep Polyclonal Antibody (Biotin)
Catalog # 103224-796
Supplier:  Novus Biologicals
Anti-FABP3 Sheep Polyclonal Antibody (Biotin)
Catalog # 103224-796
Supplier:  Novus Biologicals
Supplier Number:  BAF1678

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Fatty Acid Binding Protein 3, Muscle And Heart
  • Antigen Symbol:
    FABP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Sheep
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 ug
  • Antigen Synonyms:
    O-FABP|FABP11|MDGI|H-FABP|M-FABP
  • Storage Buffer:
    Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Immunogen:
    E. coli-derived recombinant human FABP3 Met1-Ala133 Accession # P05413
  • Purification:
    Antigen Affinity-purified
  • Cat. No.:
    103224-796
  • Supplier no.:
    BAF1678

Specifications

About this item

Type: Primary Antibodies
Antigen: FABP3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human