Anti-SOST Goat Polyclonal Antibody (Biotin)
Catalog # 103224-622
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Sclerostin
- Antigen Symbol:SOST
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:50 ug
- Antigen Synonyms:VBCH|CDD|Sclerostin|UNQ2976/PRO7455/PRO7476|SOST|SOST1
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Mouse myeloma cell line NS0-derived recombinant human SOST (R&D Systems, Catalog # 1406-ST) Gln24-Tyr213 Accession # Q9BQB4
- Purification:Antigen Affinity-purified
- Cat. No.:103224-622
- Supplier no.:BAF1406
Specifications
About this item
Type: Primary Antibodies
Antigen: SOST
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human