Anti-CCL5 Goat Polyclonal Antibody (Biotin)
Catalog # 103224-208
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:CCL5/RANTES
- Antigen Symbol:CCL5
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Goat
- Isotype:IgG
- Reactivity:Cotton rat
- Size:50 ug
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:E. coli-derived recombinant cotton rat CCL5/RANTES (R&D Systems, Catalog # 1010-CR) Ser24-Lys91 Accession # AAL16932
- Purification:Antigen Affinity-purified
- Cat. No.:103224-208
- Supplier no.:BAF1010
Specifications
About this item
Type: Primary Antibodies
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Cotton Rat