Order Entry
United States
ContactUsLinkComponent
 
Anti-APP Goat Polyclonal Antibody
  103223-892
Anti-APP Goat Polyclonal Antibody
  103223-892
 :  AF890-SP

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Amyloid beta (A4) Precursor Protein
  • Antigen Symbol:
    APP
  • Clonality:
    Polyclonal
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    25 ug
  • Antigen Synonyms:
    CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
  • Storage Buffer:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Immunogen:
    S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP695+1 frameshift mutant Met1-Arg336 Accession # NP_958817
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103223-892
  • Supplier no.:
    AF890-SP

 

 

Type: Primary Antibodies
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human