Order Entry
United States
ContactUsLinkComponent
Anti-TYRO3 Goat Polyclonal Antibody
Anti-TYRO3 Goat Polyclonal Antibody
Catalog # 103223-798
Supplier:  Novus Biologicals
Anti-TYRO3 Goat Polyclonal Antibody
Catalog # 103223-798
Supplier:  Novus Biologicals
Supplier Number:  AF859

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tyro3 Protein Tyrosine Kinase
  • Antigen Symbol:
    TYRO3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 ug
  • Antigen Synonyms:
    Sky|etk2/tyro3|tk19-1|TK19-2|Rse|Tif|AI323366|Dtk|Brt|Etk-2
  • Storage Buffer:
    Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
  • Storage Temperature:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Immunogen:
    S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dtk Ala41-Ser428 Accession # Q06418
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    103223-798
  • Supplier no.:
    AF859

Specifications

About this item

Type: Primary Antibodies
Antigen: TYRO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human