Anti-PEAR1 Sheep Polyclonal Antibody
103222-062
- Antibody Type:Primary
- Antigen Name:Platelet Endothelial Aggregation Receptor 1
- Antigen Symbol:PEAR1
- Clonality:Polyclonal
- Host:Sheep
- Isotype:IgG
- Reactivity:Mouse
- Size:100 ug
- Antigen Synonyms:PEAR1|MEGF12|Platelet endothelial aggregation receptor 1|hPEAR1|Multiple epidermal growth factor-like domains protein 12|JEDI
- Storage Buffer:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
- Immunogen:Chinese hamster ovary cell line CHO-derived recombinant mouse PEAR1 Leu19-Leu754 Accession # Q8VIK5
- Purification:Immunogen affinity purified
- Cat. No.:103222-062
- Supplier no.:AF7607
Type: Primary Antibodies
Antigen: PEAR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse