Diamonds Medium, Modified, for Trichomonas, Hardy Diagnostics
Catalog # 10324-450
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass tube
- Description:Diamonds Medium, Modified, for Trichomonas
- Size:6 ml
- Cat. No.:10324-450
- Tube size:16×100 mm
- Supplier no.:K02
Specifications
About this item
Selective cultivation of Trichomonas spp from clinical specimens.
- Selective Trichmonas cultivation
- Clinical specimen cultivation
For the selective cultivation of Trichomonas spp., especially T. vaginalis from clinical specimens.