About this item
Soft cloth backing is breathable, flexible and conformable, yet is gentle to the skin for increased patient comfort.
- Solid gel conforms to the skin quickly allowing high quality, uninterrupted traces
- Low chloride gel content minimizes skin irritation for increased patient comfort
- Provides good performance for up to 30 days out of the bag
Designed for use in ECG monitoring applications.