About this item
Multi-purpose monitoring electrodes with sticky gel feature a high performance adhesive and foam backing.
- Foam backing is comfortable for patients, occlusive to fluids, and easy to handle for quick application
- Sticky gel delivers excellent trace quality resulting in better patient information
- Larger adhesive and gel areas to improve product performance
They are clinically proven to stay in place for up to five days. Ideally suited for OR, ER, and diagnostic stress testing applications.