Order Entry
United States
ContactUsLinkComponent
Tryptic Soy Broth, General Laboratory Products
Tryptic Soy Broth, General Laboratory Products
Catalog # 101205-572
Minority or Woman-Owned Business Enterprise
CAS Number:  
Tryptic Soy Broth, General Laboratory Products
Catalog # 101205-572
Supplier Number:  16T-3800-10
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Tryptic Soy Broth, 6 x 125 mm Polycarbonate Tube
  • Size:
    10 mL
  • Cat. No.:
    101205-572
  • Supplier no.:
    16T-3800-10

Specifications

About this item

Base for a general purpose medium.