About this item
Ready-to-use, conductive gel pads for use in defibrillation.
- Bright orange solid gel pads are easy to see
- Require no cleanup, and leave no slippery residue that can interfere with CPR
Self-contained to eliminate inadvertent gel bridging which can cause skin trauma to patient or operator.