ContactUsLinkComponent
Curos Tips™ Disinfecting Cap Strip for Male Luers, 3M™
Curos Tips™ Disinfecting Cap Strip for Male Luers, 3M™
Catalog # 75788-304
Curos Tips™ Disinfecting Cap Strip for Male Luers, 3M™
Catalog # 75788-304
Supplier Number:  MMM CM5-200
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Curos Tips™ Disinfecting Cap Strip for Male Luers
  • Cat. No.:
    75788-304
  • Supplier no.:
    MMMCM5-200
Specifications
About this item
Curos Tips™ use 70% isopropyl alcohol (IPA) to passively disinfect and protect all critical surfaces of male luer connections on intravenous tubing sets.
  • Helps reduce the risk of contaminants from entering the catheter post-insertion
  • Fits majority of male luer connections
  • Disinfects critical surfaces of male luer connections in 1 minute
  • Packaged to hang from I.V. pole for easy access, greater compliance, and reduced waste