ContactUsLinkComponent
Curos™ Disinfecting Cap for Tego® Hemodialysis Connectors, 3M™
Catalog # 75788-036
Curos™ Disinfecting Cap for Tego® Hemodialysis Connectors, 3M™
Catalog # 75788-036
Supplier Number:  MMM CTG1-270
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Curos™ Disinfecting Cap for Tego® Hemodialysis Connectors
  • Cat. No.:
    75788-036
  • Supplier no.:
    MMMCTG1-270
Specifications
About this item
A disinfecting port protector approved for the Tego® needlefree hemodialysis connector.
  • Helps reduce the risk of contaminants from entering the catheter post-insertion
  • Disinfects in 3 minutes
These white Curos™ caps are easily distinguished for dedicated use on the Tego® connectors, and are designed specifically to maintain the integrity of the Tego® valve’s surface.