ContactUsLinkComponent
Cavilon™ Durable Barrier Cream, 3M™
Cavilon™ Durable Barrier Cream, 3M™
Catalog # 75786-412
Cavilon™ Durable Barrier Cream, 3M™
Catalog # 75786-412
Supplier Number:  MMM 3355
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Capacity:
    96 ml (3.25 oz.)
  • Description:
    Cavilon™ Durable Barrier Cream
  • Cat. No.:
    75786-412
  • Supplier no.:
    MMM3355
Specifications
About this item
A concentrated cream that provides durable, long-lasting protection from body fluids while moisturizing the skin
  • Hypoallergenic
  • CHG compatible
  • Contains Dimethicone for skin protection
  • Fragrance-free
Unique, polymer-based formula provides a breathable barrier to promote skin health in patients at risk for skin damage from incontinence. The cream vanishes into the skin, does not clog briefs or diapers, and resists wash off. Using Cavilon Durable Barrier Cream regularly on intact skin can help prevent breakdown caused by regular exposure to urine and stool.