Specifications
- Description:Model 585 Lithotomy Warming Blanket, with 61 x 61 cm (24 x 24") Drape
- Size:188 x 91.4 cm (74 x 36")
- Cat. No.:75787-606
- Supplier no.:MMM58501
Specifications
About this item
This advanced blanket delivers full, unrestricted patient access and flexibility for procedures involving the lower extremities and abdominal, peritoneal, and pelvic cavities.
- Unique fluid outlets minimize pooling of fluids on the blanket surface
- Pass-through slits allow flexible patient positioning and the use of a drawsheet
- Integrated tie strips can be used to secure the blanket to the stirrups/leg supports
- Soft, radiolucent materials accommodate imaging requirements
- Adhesive strips and tuck flaps secure the blanket to the OR/procedure table
Generous peritoneal cut-out at the base of the blanket provides the clinician unobstructed access to the patient. One clear plastic drape (included) helps retain warm air around the head of the patient.