ContactUsLinkComponent
Thomas-Gaylor Biopsy Forceps, OR Grade, Sklar
Thomas-Gaylor Biopsy Forceps, OR Grade, Sklar
Catalog # 10198-798
Supplier:  Sklar
undefined
Thomas-Gaylor Biopsy Forceps, OR Grade, Sklar
Catalog # 10198-798
Supplier:  Sklar
Supplier Number:  80-1695
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    241 mm (9½")
  • Material:
    Stainless steel
  • Tip configuration:
    Interfitted Round Cups
  • Curvature:
    Double ('S') Curved
  • Cat. No.:
    10198-798
  • Supplier no.:
    80-1695
Specifications
About this item
Thomas-Gaylor Biopsy Forceps are constructed of stainless steel
Industry standard.