Order Entry
United States
ContactUsLinkComponent
 
Wilmer Iris Scissors, OR Grade, Sklar
  10197-862
 :  Sklar
undefined
Wilmer Iris Scissors, OR Grade, Sklar
  10197-862
 :  Sklar
 :  64-1735

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Length:
    102 mm (4")
  • Material:
    Stainless steel
  • Curvature:
    Angled
  • Cat. No.:
    10197-862
  • Supplier no.:
    64-1735
  • Blade type:
    Sharp/Sharp

 

 

The Wilmer Iris Scissors, also known as Wilmer Conjunctival Scissors or Wilmer (Converse) Scissors, are most commonly used during ophthalmological procedures for lifting the conjunctival flap

The 45°, angled, dual sharp tips also make this instrument useful for trimming nasal cartilage.