ContactUsLinkComponent
ULTRA-WARE® Standard Five Valve Recirculation/Filtration Cap, Kimble Chase, DWK Life Sciences
ULTRA-WARE® Standard Five Valve Recirculation/Filtration Cap, Kimble Chase, DWK Life Sciences
Catalog # KT953983-0047
undefined
ULTRA-WARE® Standard Five Valve Recirculation/Filtration Cap, Kimble Chase, DWK Life Sciences
Catalog # KT953983-0047
Supplier Number:  953983-0047
Specifications
  • Cap Size:
    GL 45
  • Ø:
    47 mm
  • Description:
    Standard Five Valve Recirculation/Filtration Cap
  • Environmentally Preferable:
  • Cat. No.:
    KT953983-0047
  • Supplier no.:
    953983-0047
Specifications
About this item
Filtration cap designed to permit the recirculation of mobile phase back into the reservoir while maintaining the helium sparged environment.
  • Performs stepwise filtration, sparging/degassing, storage and delivery to the HPLC pump
  • Integral check valve prevents mobile phase from backing up into the gas lines
  • Pressure release valve protects the reservoir from being over-pressurized
  • 1/4"-28 fitting connections allow easy connection to any HPLC pump system
  • Cap body made from glass-filled PTFE fits any reservoir or bottle with a GL 45 thread