PAD Etch 4
Catalog # 89400-808
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle
- Size:1 gal.
- Cat. No.:89400-808
- Supplier no.:444-064251
Specifications
Specification Test Results
- Assay (CH3COOH):31.00 - 32.00%
- Assay (NH4F):12.80 - 13.80%
- Color:10
- Appearance:Conform
- Aluminum:3000 ppb
- Antimony:1000 ppb
- Arsenic:1000 ppb
- Barium:1000 ppb
- Boron:300 ppb
- Cadmium:500 ppb
- Calcium:500 ppb
- Chromium:500 ppb
- Cobalt:500 ppb
- Copper:1000 ppb
- Gallium:500 ppb
- Germanium:500 ppb
- Gold:500 ppb
- Iron:1000 ppb
- Lead:1000 ppb
- Lithium:1000 ppb
- Magnesium:500 ppb
- Manganese:500 ppb
- Nickel:500 ppb
- Potassium:2000 ppb
- Silver:500 ppb
- Strontium:1000 ppb
- Tin:500 ppb
- Titanium:500 ppb
- Zinc:500 ppb
Specification Test Results
About this item
: