ContactUsLinkComponent
Arizant Bair Hugger™ Cardiac Warming Blanket, 3M™
  75786-392
Arizant Bair Hugger™ Cardiac Warming Blanket, 3M™
  75786-392
 :  MMM 63000
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Model 630 Cardiac Warming Blanket
  • Size:
    152.4 x 91.4 cm (60 x 36")
  • Environmentally Preferable:
  • Cat. No.:
    75786-392
  • Supplier no.:
    MMM63000
This sterile blanket is perfect for warming the lower body during cardiothoracic and vascular procedures.
  • Warms the lower body to reduce core-to-peripheral heat loss
  • Positions easily in the sterile field
  • Clear groin/femoral access window allows access to the femoral artery
  • Soft, radiolucent materials will not interfere with imaging requirements
  • Foot drape minimizes risk of thermal injury to the feet and lower leg area
  • 36" sterile hose allows the placement of the warming unit away from sterile field