ContactUsLinkComponent
Arizant Bair Hugger™ Adult Underbody Warming Blanket, 3M™
Arizant Bair Hugger™ Adult Underbody Warming Blanket, 3M™
Catalog # 75787-598
Arizant Bair Hugger™ Adult Underbody Warming Blanket, 3M™
Catalog # 75787-598
Supplier Number:  MMM 54500
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Model 545 Adult Underbody Warming Blanket
  • Size:
    188 x 91.4 cm (74 x 36")
  • Environmentally Preferable:
  • Cat. No.:
    75787-598
  • Supplier no.:
    MMM54500
Specifications
About this item
This underbody blanket provides full, unrestricted patient access and is conveniently positioned on the procedure table before the patient arrives, so it's ready when you are.
  • Underbody design warms the patient while providing full, unrestricted patient access
  • Ideal for patient warming during diagnostic and interventional procedures
  • Unique fluid outlets minimize pooling of fluids on the blanket surface
  • Consistent, even perforations across the entire blanket ensure uniform convective warming
  • Soft, radiolucent materials do not interfere with imaging
  • Two adhesive strips under the blanket secure the it to the OR/procedure table