ContactUsLinkComponent
Round Bottom Flasks, Minum-ware, MW-58, Chemglass
Round Bottom Flasks, Minum-ware, MW-58, Chemglass
Catalog # 80064-168
Supplier:  Chemglass
Small Business Enterprise
undefined
Round Bottom Flasks, Minum-ware, MW-58, Chemglass
Catalog # 80064-168
Supplier:  Chemglass
Supplier Number:  MW-58-02
Small Business Enterprise
Specifications
  • [ST] joint no.:
    14/10
  • Capacity:
    10 ml
  • Description:
    Round Bottom Flask, Single Neck
  • Cat. No.:
    80064-168
  • Supplier no.:
    MW-58-02
Specifications
About this item
Micro round bottom flask with a 14/10 threaded outer joint
Supplied complete with an open-top screw cap and Viton O-ring.