Ammonium fluoride 40% in aqueous solution MB
89400-748
: KMG
:
- Pack type:Poly bottle
- Size:1 gal.
- Cat. No.:89400-748
- Supplier no.:404-063915
- Assay (NH4F):40.00 - 41.00
- Color:10
- Appearance:Conform
- pH of 1% Solution:6.00 - 7.00
- Chloride:2000 ppb
- Nitrate:3000 ppb
- Phosphate:400 ppb
- Sulfate:1000 ppb
- Aluminum:10 ppb
- Antimony:5 ppb
- Arsenic:5 ppb
- Barium:2 ppb
- Beryllium:1 ppb
- Bismuth:1 ppb
- Boron:5 ppb
- Cadmium:1 ppb
- Calcium:10 ppb
- Chromium:3 ppb
- Cobalt:1 ppb
- Copper:1 ppb
- Gallium:1 ppb
- Germanium:5 ppb
- Gold:5 ppb
- Iron:10 ppb
- Lead:5 ppb
- Lithium:1 ppb
- Magnesium:5 ppb
- Manganese:1 ppb
- Molybdenum:1 ppb
- Nickel:5 ppb
- Niobium:1 ppb
- Potassium:6 ppb
- Silver:1 ppb
- Sodium:7 ppb
- Strontium:1 ppb
- Tantalum:3 ppb
- Thallium:5 ppb
- Tin:5 ppb
- Titanium:1 ppb
- Vanadium:1 ppb
- Zinc:5 ppb
- Zirconium:1 ppb
- 0.20µ Particle Count:1500 par/ml
- 0.3µ Particle Count:1500 par/ml
- 0.5µ Particle Count:50 par/ml
- 1.0µ Particle Count:10par/ml
:
:37.04 g/mol
:3287
:6.1,III
:12125-01-8
:NH₄F
:MFCD00011423