TransIT-TKO® Transfection Reagent, Mirus Bio
Supplier: Mirus Bio
Certificates
About this item
A high-efficiency, low-toxicity siRNA transfection reagent for mammalian cells.
- Broad spectrum siRNA and plasmid DNA delivery
- Low cellular toxicity
- Reproducible results
- High knockdown efficiency
Maintain cell density and reduce experimental biases with this reagent. One transfection reagent and protocol can be utilized for a wide variety of cells. Achieve optimal gene silencing in a large percentage of cells to ensure experimental success.
Specifications
- siRNA:Yes