TransIT®-QR Delivery Solution and Starter Kit, Mirus Bio
Catalog # 10767-006
Supplier: Mirus Bio
CAS Number:
Specifications
- DNA and siRNA:Yes
- DNA:Yes
- In Vivo:Yes
- siRNA:Yes
- Description:TransIT®-QR Delivery Solution and Starter Kit
- Size:40 injections
- Cat. No.:10767-006
- Supplier no.:MIR5240
Specifications
About this item
A nontoxic, "Quick Recovery" in vivo delivery solution that delivers DNA or siRNA to the livers of laboratory mice using hydrodynamic tail vein injection.
- Naked nucleic acid delivery
- Versatile platform
- Low toxicity
Each TransIT®-QR Starter Kit contains the TransIT®-QR Delivery Solution, 10 syringes, needles and alcohol swabs, and a conical plastic tube to restrain the mouse while tail vein injection is performed. The TransIT®-QR Delivery Solution is also sold separately as indicated above.