TransIT®-EE Delivery Solution and Starter Kit, Mirus Bio
Catalog # 10767-012
Supplier: Mirus Bio
CAS Number:
Specifications
- DNA:Yes
- In Vivo:Yes
- Description:TransIT®-EE Delivery Solution and Starter Kit
- Size:40 injections
- Cat. No.:10767-012
- Supplier no.:MIR5340
Specifications
About this item
This solution and starter kit is an "enhanced expression" in vivo delivery solution that delivers DNA to the livers of laboratory mice using hydrodynamic tail vein injection.
- Naked nucleic acid delivery
- Versatile platform
Achieve increased levels of gene expression than compared to TransIT®-QR Delivery Solution, TransIT- In Vivo Gene Delivery System, polylysine and PEI methods. This solution is suitable for DNA delivery using hydrodynamic tail vein injection to the mouse strain of your choice.