TransIT®-Jurkat Transfection Reagent, Mirus Bio
A high-efficiency DNA transfection reagent optimized for Jurkat and other hard-to-transfect cell types.
- Unique formulation
- High-efficiency delivery
- Deliver single or multiple plasmids
Maximize transfection performance in Jurkat cells and other hard-to-transfect cells such as K562, RAW 264.7, and THP-1 cells.
Reagent is suitable for many applications such as gene expression and promoter analysis.
- Cell Specific:Jurkat
- DNA:Yes
Frequently Bought Together