TransIT-HeLaMONSTER® Transfection Kit, Mirus Bio
A high-efficiency, low-toxicity DNA transfection reagent optimized for HeLa cells.
- Unique formulation
- Low cellular toxicity
- High-efficiency delivery
- Deliver single or multiple plasmids
Achieve a 50-60% transfection efficiency with the use of these kits.
Each kit contains a base and booster reagent to maximize transfection performance in HeLa cells.
- Environmentally Preferable:
Frequently Bought Together