Plastic Backed Student Thermometer, United Scientific Supplies
Catalog # 470222-098
Supplier: United Scientific Supplies
Specifications
- Cat. No.:470222-098
- Range:−40…+50 °C
- Supplier no.:THMPB1-PK/15
Specifications
About this item
Great for classroom use.
- Glass thermometer mounted on plastic backing card
- Filled with kerosene solution
- Celsius scale only
This large scale thermometer is perfect for being seen anywhere in a room. Only has Celsius scale.