Metal Backed Student Dual Scale Thermometer, United Scientific Supplies
470222-094
- Cat. No.:470222-094
- Range:−20…+110 °C (0...230 °F)
- Supplier no.:THBV02
Great for classroom use.
- Glass thermometer mounted on aluminum back
- Filled with kerosene solution
- Celsius and Fahrenheit scales
Glass thermometer mounted on aluminum back. Thermometers are filled with approximately 3 grams of kerosene mixed with red color.