Anti-LMNA Mouse Monoclonal Antibody [clone: GT9712]
Catalog # 10167-388
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Lamin A/C
- Antigen Symbol:LMNA
- Clonality:Monoclonal
- Clone:GT9712
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:4000
- Antigen Synonyms:LMNL1|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|LFP|FPLD2|LDP1|CDDC|CMD1A|EMD2|HGPS
- Storage Buffer:PBS
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human Lamin A/C. The exact sequence is proprietary.
- Purification:Affinity purified by Protein G.
- Cat. No.:10167-388
- Supplier no.:GTX629404
Specifications
About this item
Mouse monoclonal antibody [GT9712] to Lamin A + C
Type: Primary
Antigen: LMNA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human