Order Entry
United States
ContactUsLinkComponent
Anti-SCGB1A1 Mouse Monoclonal Antibody [clone: 3A8B8]
Anti-SCGB1A1 Mouse Monoclonal Antibody [clone: 3A8B8]
Catalog # 10169-208
Supplier:  Genetex
Anti-SCGB1A1 Mouse Monoclonal Antibody [clone: 3A8B8]
Catalog # 10169-208
Supplier:  Genetex
Supplier Number:  GTX60655

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Secretoglobin, Family 1A, Member 1
  • Antigen Symbol:
    SCGB1A1
  • Clonality:
    Monoclonal
  • Clone:
    3A8B8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Gene ID:
    7356
  • Antigen Synonyms:
    UP-1|Uteroglobin|UGB|CC10|CC16|UP1|CCPBP|CCSP
  • Storage Buffer:
    Purified antibody in PBS with 0.05% sodium azide and 0.5% protein stabilizer.
  • Storage Temperature:
    4 °C; –20 °C for long term storage
  • Concentration:
    1 mg/ml
  • Immunogen:
    The immunogen corresponding to a region within amino acids 26 and 91 of human SCGB1A1 expressed in E. Coli.
  • Cat. No.:
    10169-208
  • Supplier no.:
    GTX60655

Specifications

About this item

Mouse Monoclonal Antibody [3A8B8] to SCGB1A1

Type: Primary
Antigen: SCGB1A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human