Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
10168-894
: Genetex
- Antibody Type:Primary
- Antigen Name:Protein Kinase AMP-activated Alpha 1 Catalytic Subunit
- Antigen Symbol:PRKAA1
- Clonality:Monoclonal
- Clone:2B7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:5562
- Antigen Synonyms:catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:4 °C; –20 °C for long term storage
- Immunogen:The immunogen corresponding to human PRKAA1 expressed in E. Coli.
- Cat. No.:10168-894
- Supplier no.:GTX60403
Mouse monoclonal antibody [2B7] to AMPK alpha 1
Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Rat