Order Entry
United States
ContactUsLinkComponent
Anti-MAP2K6 Mouse Monoclonal Antibody [clone: 3H12B9]
Anti-MAP2K6 Mouse Monoclonal Antibody [clone: 3H12B9]
Catalog # 10168-172
Supplier:  Genetex
Anti-MAP2K6 Mouse Monoclonal Antibody [clone: 3H12B9]
Catalog # 10168-172
Supplier:  Genetex
Supplier Number:  GTX60397
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Mitogen-Activated Protein Kinase Kinase 6
  • Antigen Symbol:
    MAP2K6
  • Clonality:
    Monoclonal
  • Clone:
    3H12B9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    5608
  • Antigen Synonyms:
    PRKMK6|MKK6|MAPKK6|SAPKK-3|MEK6|SAPKK3
  • Storage Buffer:
    Ascitic fluid containing 0.03% sodium azide.
  • Storage Temperature:
    4 °C; –20 °C for long term storage
  • Immunogen:
    The immunogen corresponding to human MAP2K6 expressed in E. Coli.
  • Cat. No.:
    10168-172
  • Supplier no.:
    GTX60397

Specifications

About this item

Mouse Monoclonal Antibody [3H12B9] to MAP2K6

Type: Primary
Antigen: MAP2K6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse