Anti-SOAT2 Rabbit Polyclonal Antibody
Catalog # 10165-112
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Sterol O-Acyltransferase 2
- Antigen Symbol:SOAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 μg
- Gene ID:8435
- Antigen Synonyms:ACACT2|ARGP2|ACAT2
- Storage Buffer:100ug/ml BSA, 50% glycerol and 0.09% sodium azide.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:KLH conjugated synthetic peptide derived from human ACAT2 (355-400aa)
- Purification:Purified by Protein A and peptide affinity chromatography.
- Cat. No.:10165-112
- Supplier no.:GTX60244
Specifications
About this item
Rabbit Polyclonal antibody to SOAT2
Type: Primary
Antigen: SOAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat